Learn More
Waters Corp Waters Corporation VanGuard Cartridge, XBridge BEH Phenyl, 130Å, 5 μm, 2.1 mm × 5 mm
Catalog No. 502038207 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream. This cartridge is specifically optimized to protect all 2.1 mm and 3.0 mm inner diameter XBridge BEH Phenyl analytical columns that contain 5 μm sorbent particles.
  • Designed to extend the lifetime and performance of analytical columns.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 2.1 mm and 3.0 mm inner diameter analytical columns.
  • Contains 5 μm sorbent particles.
Catalog No. 50-203-8207 Supplier Waters Corp Supplier No. 186007796
XBridge BEH Phenyl VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream. This cartridge is specifically optimized to protect all 2.1 mm and 3.0 mm inner diameter XBridge BEH Phenyl analytical columns that contain 5 μm sorbent particles.
  • Designed to extend the lifetime and performance of analytical columns.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 2.1 mm and 3.0 mm inner diameter analytical columns.
  • Contains 5 μm sorbent particles.