Learn More
Waters Corp Waters Corporation Column, 130Å, 2.5 μm, 2.1 mm ID, 30 mm Length
Catalog No. 50179490 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases for polyaromatic compounds. Featuring a highly stable Phenyl sorbent, they provide ultra-low column bleed, long column lifetimes, and excellent peak shape. These 2.5 μm XP columns deliver exceptional separation performance and robustness for HPLC assays, supporting UPLC adoption and wider pH range technique development.
  • Complementary selectivity to C18 stationary phases.
  • Chemically stable Phenyl sorbent for ultra-low column bleed and long lifetimes.
  • Exceptional separation performance and robustness with ultra-low dispersion hardware.
  • Wider pH range and excellent low and high pH stability.
  • Increased longevity, superior reliability, and exceptional particle efficiency.
Catalog No. 50-179-490 Supplier Waters Corp Supplier No. 186006064
Add to Cart
Add to Cart
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases for polyaromatic compounds. Featuring a highly stable Phenyl sorbent, they provide ultra-low column bleed, long column lifetimes, and excellent peak shape. These 2.5 μm XP columns deliver exceptional separation performance and robustness for HPLC assays, supporting UPLC adoption and wider pH range technique development.
  • Complementary selectivity to C18 stationary phases.
  • Chemically stable Phenyl sorbent for ultra-low column bleed and long lifetimes.
  • Exceptional separation performance and robustness with ultra-low dispersion hardware.
  • Wider pH range and excellent low and high pH stability.
  • Increased longevity, superior reliability, and exceptional particle efficiency.