Learn More
Waters Corp WATERS Column, Phenyl, 2.5 μm, 2.1 mm, 50 mm
Catalog No. 50179491 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Phenyl columns offer complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. They feature the most chemically stable Phenyl sorbent, ensuring ultra-low column bleed, extended lifetimes, and excellent peak shape. Packed with 2.5 μm XP particles in ultra-low dispersion hardware, these columns provide exceptional separation performance, robustness, and throughput for HPLC assays, also facilitating UPLC adoption.
  • Offers complementary selectivity to C18 phases.
  • Features chemically stable Phenyl sorbent for extended column lifetimes.
  • 2.5 μm XP particles enable exceptional performance and throughput for HPLC assays.
  • Wide usable pH range (1 to 12) for robust development.
  • Outstanding low and high pH stability ensures high repeatability.
  • Facilitates a seamless transition to UPLC adoption.
Catalog No. 50-179-491 Supplier Waters Corp Supplier No. 186006065
Phenyl columns offer complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. They feature the most chemically stable Phenyl sorbent, ensuring ultra-low column bleed, extended lifetimes, and excellent peak shape. Packed with 2.5 μm XP particles in ultra-low dispersion hardware, these columns provide exceptional separation performance, robustness, and throughput for HPLC assays, also facilitating UPLC adoption.
  • Offers complementary selectivity to C18 phases.
  • Features chemically stable Phenyl sorbent for extended column lifetimes.
  • 2.5 μm XP particles enable exceptional performance and throughput for HPLC assays.
  • Wide usable pH range (1 to 12) for robust development.
  • Outstanding low and high pH stability ensures high repeatability.
  • Facilitates a seamless transition to UPLC adoption.