Learn More
Waters Corp XBridge Column, BEH Phenyl, 2.5 μm, 4.6 x 30 mm
Catalog No. 50201526 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. They are chemically stable phenyl sorbents, providing ultra-low column bleed, extended lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, the 2.5 μm XP columns deliver exceptional separation performance and robustness for HPLC assays, facilitating UPLC adoption.
  • Unparalleled column longevity for chromatographic analysis.
  • Wide operating range for mobile phase pH (1 to 12) and temperature.
  • Facilitates seamless UPLC technology conversion.
  • Provides alternative selectivity for analytes with aromatic rings.
  • BEH particle technology ensures robust technique development and pH stability.
  • Withstands a maximum pressure of 10,000 psi (690 Bar).
Catalog No. 50-201-526 Supplier Waters Corp Supplier No. 186006072
These XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. They are chemically stable phenyl sorbents, providing ultra-low column bleed, extended lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, the 2.5 μm XP columns deliver exceptional separation performance and robustness for HPLC assays, facilitating UPLC adoption.
  • Unparalleled column longevity for chromatographic analysis.
  • Wide operating range for mobile phase pH (1 to 12) and temperature.
  • Facilitates seamless UPLC technology conversion.
  • Provides alternative selectivity for analytes with aromatic rings.
  • BEH particle technology ensures robust technique development and pH stability.
  • Withstands a maximum pressure of 10,000 psi (690 Bar).