Learn More
Waters Corp Waters Column, BEH Phenyl, 2.5 um Particle, 130 A Pore
Catalog No. 50179230 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. This product features the most chemically stable commercially available Phenyl sorbent, providing ultra-low column bleed, long column lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a seamless transition to UPLC adoption.
  • Complementary selectivity to C18 stationary phases
  • Chemically stable Phenyl sorbent with ultra-low column bleed
  • Extended column lifetimes and excellent peak shape
  • Packed in ultra-low dispersion hardware
  • 2.5 μm XP technology for exceptional separation, robustness, and high throughput in HPLC assays
  • Seamless transition to UPLC adoption
Catalog No. 50-179-230 Supplier Waters Corp Supplier No. 176002571
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. This product features the most chemically stable commercially available Phenyl sorbent, providing ultra-low column bleed, long column lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, while also enabling a seamless transition to UPLC adoption.
  • Complementary selectivity to C18 stationary phases
  • Chemically stable Phenyl sorbent with ultra-low column bleed
  • Extended column lifetimes and excellent peak shape
  • Packed in ultra-low dispersion hardware
  • 2.5 μm XP technology for exceptional separation, robustness, and high throughput in HPLC assays
  • Seamless transition to UPLC adoption