Learn More
Waters Corp Waters Column, BEH Phenyl, 2.5 μm, 2.1 x 75 mm
Catalog No. 50179231 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
eXtended Performance 2.5 μm columns facilitate the transition between HPLC and UPLC™ technology. Compatible with any LC system, they offer flexibility for HPLC, UHPLC, and UPLC users, improving productivity at intermediate backpressures. Diverse stationary phases enable seamless scalability across platforms.
  • Extending separation performance
  • Compatible with any LC system (HPLC, UHPLC and UPLC)
  • Improved productivity at intermediate backpressures
  • Diverse range of stationary phases
  • Directly scalable to HPLC and UPLC columns
  • Ultra-high pressure compatible with 40% lower backpressure
  • Superior efficiency and mass transfer
  • Higher throughput and rapid method optimization
  • Superior loading capacity for complex mixtures
Catalog No. 50-179-231 Supplier Waters Corp Supplier No. 176002572
eXtended Performance 2.5 μm columns facilitate the transition between HPLC and UPLC™ technology. Compatible with any LC system, they offer flexibility for HPLC, UHPLC, and UPLC users, improving productivity at intermediate backpressures. Diverse stationary phases enable seamless scalability across platforms.
  • Extending separation performance
  • Compatible with any LC system (HPLC, UHPLC and UPLC)
  • Improved productivity at intermediate backpressures
  • Diverse range of stationary phases
  • Directly scalable to HPLC and UPLC columns
  • Ultra-high pressure compatible with 40% lower backpressure
  • Superior efficiency and mass transfer
  • Higher throughput and rapid method optimization
  • Superior loading capacity for complex mixtures