Learn More
Waters Corp Waters Phenyl Column, 130Å, 2.5 μm, 4.6 mm × 75 mm
Supplier:  Waters Corp 186006074
Encompass_Preferred
These Phenyl columns provide complementary selectivity to C18 phases, featuring a chemically stable phenyl sorbent for ultra-low column bleed, long lifetimes, and excellent peak shape. The 2.5 μm XP columns are packed in ultra-low dispersion hardware, delivering exceptional HPLC/UPLC performance, robustness, and a wider pH range for versatile method development.
  • Complementary C18 selectivity for polyaromatic compounds.
  • Chemically stable phenyl sorbent; ultra-low column bleed.
  • Long column lifetimes; excellent peak shape.
  • 2.5 μm XP columns in ultra-low dispersion hardware.
  • Exceptional HPLC/UPLC separation performance and throughput.
  • Wider pH range; robust method development.
  • Trifunctional bonding for high stability.
  • Improved longevity, reliability, particle efficiency.
Catalog No. 50-201-528