Learn More
Waters Corp Waters, XBridge BEH Phenyl XP Column, 130 Å, 2.5 μm, 3 mm X 100 mm, 3/pk
Catalog No. 50179236 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. Featuring the most chemically stable Phenyl sorbent, they offer ultra-low column bleed, extended lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC adoption.
  • Phenyl chemistry and reversed phase separation mode.
  • Hybrid particle substrate, spherical shape.
  • pH range of 1 to 12.
  • Maximum pressure of 18000 psi (1240 bar).
  • Endcapped with phenyl-hexyl bonding technology, low silanol activity.
  • Pore size: 130 Å; dimensions: 3 mm × 100 mm.
  • 15% carbon load.
  • eCord enabled, 3 units per package.
Catalog No. 50-179-236 Supplier Waters Corp Supplier No. 176002577
XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. Featuring the most chemically stable Phenyl sorbent, they offer ultra-low column bleed, extended lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC adoption.
  • Phenyl chemistry and reversed phase separation mode.
  • Hybrid particle substrate, spherical shape.
  • pH range of 1 to 12.
  • Maximum pressure of 18000 psi (1240 bar).
  • Endcapped with phenyl-hexyl bonding technology, low silanol activity.
  • Pore size: 130 Å; dimensions: 3 mm × 100 mm.
  • 15% carbon load.
  • eCord enabled, 3 units per package.