Learn More
Waters Corp Waters Corporation XBridge BEH Phenyl XP Column, 130Å, 2.5 μm, 3 mm X 100 mm
Catalog No. 50179497 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Phenyl XP Column provides complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. It is a chemically stable phenyl sorbent, ensuring ultra-low column bleed, extended lifetimes, and excellent peak shape for robust and efficient HPLC assays, also supporting UPLC adoption.
  • Most chemically stable phenyl sorbent, ensuring ultra-low column bleed and long lifetimes
  • Provides excellent peak shape and exceptional separation performance
  • 2.5 μm eXtended performance (XP) columns with ultra-low dispersion hardware
  • Enables robustness and high throughput for HPLC assays and UPLC adoption
  • Offers wide pH range for method development due to BEH particle technology
  • Trifunctional bonding of phenyl ligand ensures high pH stability
  • Provides alternative selectivity for analytes with aromatic rings, aiding challenging separations
Catalog No. 50-179-497 Supplier Waters Corp Supplier No. 186006071
The XBridge BEH Phenyl XP Column provides complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. It is a chemically stable phenyl sorbent, ensuring ultra-low column bleed, extended lifetimes, and excellent peak shape for robust and efficient HPLC assays, also supporting UPLC adoption.
  • Most chemically stable phenyl sorbent, ensuring ultra-low column bleed and long lifetimes
  • Provides excellent peak shape and exceptional separation performance
  • 2.5 μm eXtended performance (XP) columns with ultra-low dispersion hardware
  • Enables robustness and high throughput for HPLC assays and UPLC adoption
  • Offers wide pH range for method development due to BEH particle technology
  • Trifunctional bonding of phenyl ligand ensures high pH stability
  • Provides alternative selectivity for analytes with aromatic rings, aiding challenging separations