Learn More
Waters Corp VanGuard Cartridge, XBridge BEH Phenyl XP, 130 Å, 2.5 μm, 2.1 mm X 5 mm
Catalog No. 502390806 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination, maintaining UPLC, UHPLC, and HPLC separation efficiency.
  • Optimized to protect all 2.1 mm and 3.0 mm I.D. XBridge BEH Phenyl analytical columns with 2.5 μm sorbent particles.
  • Ensures longevity of UPLC XBridge BEH Phenyl analytical columns without compromising chromatographic performance.
  • Designed for reversed-phase liquid chromatography with UPLC columns.
  • Simple to set up and operate, protecting columns from harmful pollutants.
  • Withstands pressures up to 18,000 psi (1240 bar) in applications run on ACQUITY UPLC System.
  • Features patented BEH Technology, 130 Å pore, and 2.5 μm particles.
  • Batch QC tested via peptide map for consistent performance and extended column life.
Catalog No. 50-239-0806 Supplier Waters Corp Supplier No. 186007799
XBridge BEH Phenyl VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination, maintaining UPLC, UHPLC, and HPLC separation efficiency.
  • Optimized to protect all 2.1 mm and 3.0 mm I.D. XBridge BEH Phenyl analytical columns with 2.5 μm sorbent particles.
  • Ensures longevity of UPLC XBridge BEH Phenyl analytical columns without compromising chromatographic performance.
  • Designed for reversed-phase liquid chromatography with UPLC columns.
  • Simple to set up and operate, protecting columns from harmful pollutants.
  • Withstands pressures up to 18,000 psi (1240 bar) in applications run on ACQUITY UPLC System.
  • Features patented BEH Technology, 130 Å pore, and 2.5 μm particles.
  • Batch QC tested via peptide map for consistent performance and extended column life.