Learn More
Waters Corp XBridge BEH Shield RP18 Column, 130Å, 2.5 μm, 2.1 mm X 150 mm
Catalog No. 50465923 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 columns are designed to complement XBridge BEH C18 and C8 phases, offering an important tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, particularly for phenolic compounds, compared to traditional straight-chain alkyl columns. These eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, with a 2.5 μm particle size, enable exceptional separation performance, robustness, and throughput for HPLC assays and facilitate a seamless transition to UPLC adoption.
  • Alternate selectivity, especially for phenolic compounds, due to the embedded carbamate group in the bonded phase ligand.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • Packed in ultra-low dispersion hardware.
Catalog No. 50-465-923 Supplier Waters Corp Supplier No. 176002883
Add to Cart
Add to Cart
XBridge BEH Shield RP18 columns are designed to complement XBridge BEH C18 and C8 phases, offering an important tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, particularly for phenolic compounds, compared to traditional straight-chain alkyl columns. These eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, with a 2.5 μm particle size, enable exceptional separation performance, robustness, and throughput for HPLC assays and facilitate a seamless transition to UPLC adoption.
  • Alternate selectivity, especially for phenolic compounds, due to the embedded carbamate group in the bonded phase ligand.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
  • Packed in ultra-low dispersion hardware.