Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge BEH Shield RP18 Kit, 2.5 μm, 2.1 mm x 150 mm

Catalog No. 50466465 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This method validation kit includes three XBridge BEH Shield RP18 columns, each packed with a different sorbent batch. Designed to complement XBridge BEH C18 and C8 phases, this kit is a valuable tool for HPLC method development. The embedded carbamate group offers alternate selectivity for phenolic compounds. These 2.5 μm XP columns provide exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a smooth transition to UPLC adoption. Column documents are accessible online.

  • Chemistry: C18
  • Particle size: 2.5 μm
  • Inner diameter: 2.1 mm
  • Length: 150 mm
  • pH range: 2 to 11
  • Maximum pressure: 18000 psi (1240 bar)
  • System compatibility: UPLC, UHPLC
  • Units per package: 3

Catalog No. 50-466-465 Supplier Waters Corp Supplier No. 186006763
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This method validation kit includes three XBridge BEH Shield RP18 columns, each packed with a different sorbent batch. Designed to complement XBridge BEH C18 and C8 phases, this kit is a valuable tool for HPLC method development. The embedded carbamate group offers alternate selectivity for phenolic compounds. These 2.5 μm XP columns provide exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a smooth transition to UPLC adoption. Column documents are accessible online.

  • Chemistry: C18
  • Particle size: 2.5 μm
  • Inner diameter: 2.1 mm
  • Length: 150 mm
  • pH range: 2 to 11
  • Maximum pressure: 18000 psi (1240 bar)
  • System compatibility: UPLC, UHPLC
  • Units per package: 3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.