Learn More
Waters Corp Waters XBridge BEH Shield RP18 XP Column, 2.5 μm, 3 mm X 150 mm, 130 Å
Supplier:  Waters Corp 186006716
Encompass_Preferred
XBridge BEH Shield RP18 columns provide alternate selectivity for HPLC method development, especially for phenolic compounds, and ensure exceptional separation performance for HPLC assays.
  • Compatible with pH 2 to 11.
  • Superior low pH stability; ultra-low column bleed.
  • Maximizes particle efficiency, excellent peak shape.
  • Patented Shield Technology ensures compatibility with fully aqueous mobile phases.
  • Withstands pressures up to 18000 psi (1240 Bar).
  • Extended Performance [XP] for exceptional separation.
  • Utilizes BEH particle technology.
Catalog No. 50-466-424