Learn More
Waters Corp Waters XBridge Column, 2.5 μm, 130 Å, 4.6 x 150 mm
Supplier:  Waters Corp 186006717
Encompass_Preferred
XBridge BEH Shield RP18 columns are critical tools for HPLC method development, offering alternate selectivity for phenolic compounds. These 2.5 μm eXtended Performance (XP) columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, providing a seamless transition to UPLC.
  • Complements BEH C18 and C8 phases for HPLC method development.
  • Embedded carbamate group provides alternate selectivity for phenolic compounds.
  • Exceptional separation performance, robustness, and high throughput for HPLC assays.
  • Smooth transition to UPLC adoption.
  • Operates across a wide pH range with various solvents.
  • Superior separation of bases with highly symmetrical and efficient peaks.
  • Patented Shield Technology reduces silanol interactions and improves peak shape.
  • Reduces development time and increases confidence in robust separations.
Catalog No. 50-466-425