Learn More
Waters Corp XBridge Column, BEH Shield RP18, 130 Å, 3.5 μm
Catalog No. 502392706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XBridge BEH Shield RP18 columns offer unique selectivity for HPLC method development, especially for phenolic compounds, by complementing traditional C18 and C8 phases. They incorporate an embedded carbamate group and an eConnect column tag for automatic identification and digital tracking of usage. While usable with any HPLC, these device-tagged columns provide maximum benefit when paired with the Alliance iS System and Empower software.
  • Complements C18 and C8 phases for diverse method development.
  • Embedded carbamate group yields alternative selectivity for phenolic compounds.
  • eConnect column tag allows automatic identification and digital tracing.
  • Optimized with Alliance iS System and Empower software for full benefits.
  • Wide pH operating range: 2 to 11.
  • Withstands high pressures up to 6000 psi (415 Bar).
  • Ensures consistent performance with low silanol activity.
Catalog No. 50-239-2706 Supplier Waters Corp Supplier No. 186003964RF
Waters XBridge BEH Shield RP18 columns offer unique selectivity for HPLC method development, especially for phenolic compounds, by complementing traditional C18 and C8 phases. They incorporate an embedded carbamate group and an eConnect column tag for automatic identification and digital tracking of usage. While usable with any HPLC, these device-tagged columns provide maximum benefit when paired with the Alliance iS System and Empower software.
  • Complements C18 and C8 phases for diverse method development.
  • Embedded carbamate group yields alternative selectivity for phenolic compounds.
  • eConnect column tag allows automatic identification and digital tracing.
  • Optimized with Alliance iS System and Empower software for full benefits.
  • Wide pH operating range: 2 to 11.
  • Withstands high pressures up to 6000 psi (415 Bar).
  • Ensures consistent performance with low silanol activity.