Learn More
Waters Corp Prep Column, BEH Shield RP18, 10 x 150 mm, 10 μm, 130Å
Catalog No. 502392683 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge Preparative Column is an OBD column with BEH Shield RP18 chemistry. It features dimensions of 10 x 150 mm and a particle size of 10 μm, designed for preparative chromatography applications.
  • OBD column type
  • BEH Shield RP18 chemistry
  • Dimensions of 10 x 150 mm
  • Particle size of 10 μm
  • Suitable for preparative chromatography applications
Catalog No. 50-239-2683 Supplier Waters Corp Supplier No. 186008213
This XBridge Preparative Column is an OBD column with BEH Shield RP18 chemistry. It features dimensions of 10 x 150 mm and a particle size of 10 μm, designed for preparative chromatography applications.
  • OBD column type
  • BEH Shield RP18 chemistry
  • Dimensions of 10 x 150 mm
  • Particle size of 10 μm
  • Suitable for preparative chromatography applications