Learn More
Waters Corp Waters Corporation Column, BEH Shield RP18, 2.5 μm, 3 mm x 75 mm
Supplier:  Waters Corp 186006058
Encompass_Preferred
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases, offering an important tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternative selectivity, particularly for phenolic compounds, compared to straight-chain alkyl columns. These 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware, delivering exceptional separation performance, robustness, and throughput for HPLC assays, while also providing a seamless transition path toward UPLC adoption.
  • Embedded carbamate group in the bonded phase ligand for alternate selectivity, especially for phenolic compounds.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended performance [XP] columns.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
Catalog No. 50-179-488