Learn More
Waters Corp Waters Corporation Column, Shield RP18, 2.5 μm, 3 mm x 30 mm
Supplier:  Waters Corp 186006056
Encompass_Preferred
These columns complement XBridge BEH C18 and C8 phases, offering a valuable tool for HPLC method development. They feature an embedded carbamate group for alternate selectivity, especially for phenolic compounds, compared to traditional straight chain alkyl columns. With ultra-low dispersion hardware and eXtended Performance [XP] technology, these columns deliver exceptional separation performance, robustness, throughput for HPLC assays, and a seamless transition to UPLC.
  • Complements XBridge BEH C18 and C8 phases.
  • Provides alternate selectivity for phenolic compounds.
  • Features ultra-low dispersion hardware and eXtended Performance [XP] technology.
  • Delivers exceptional separation performance, robustness, and high throughput.
  • Offers seamless transition path toward UPLC adoption.
  • Digital Certificates of Analysis (COA) available online.
Catalog No. 50-179-486