Learn More
Waters Corp Oasis HLB Plate, 60 mg/96-well, 60 μm Particle, HLB Reversed-Phase Sorbent
Catalog No. 50179482 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This hydrophilic-lipophilic balanced (HLB) reversed-phase sorbent is a unique, water-wettable polymeric sorbent. It simplifies sample preparation with a 3-step protocol, eliminating conditioning and equilibration, and allows direct loading of aqueous samples. This results in an average reduction in solvent consumption of up to 70% and a 40% savings in sample preparation time.
  • Hydrophilic-lipophilic balanced (HLB) reversed-phase sorbent
  • Water-wettable polymeric sorbent allows direct loading of aqueous samples
  • Simplified 3-step protocol for sample preparation
  • Reduces solvent consumption by up to 70%
  • Saves 40% in sample preparation time
  • Stable across a pH range of 0–14
  • Provides 60 mg/96-well format
  • Features a 60 μm particle size
  • Available in 1/pk quantity
Catalog No. 50-179-482 Supplier Waters Corp Supplier No. 186006052
Add to Cart
Add to Cart
This hydrophilic-lipophilic balanced (HLB) reversed-phase sorbent is a unique, water-wettable polymeric sorbent. It simplifies sample preparation with a 3-step protocol, eliminating conditioning and equilibration, and allows direct loading of aqueous samples. This results in an average reduction in solvent consumption of up to 70% and a 40% savings in sample preparation time.
  • Hydrophilic-lipophilic balanced (HLB) reversed-phase sorbent
  • Water-wettable polymeric sorbent allows direct loading of aqueous samples
  • Simplified 3-step protocol for sample preparation
  • Reduces solvent consumption by up to 70%
  • Saves 40% in sample preparation time
  • Stable across a pH range of 0–14
  • Provides 60 mg/96-well format
  • Features a 60 μm particle size
  • Available in 1/pk quantity