Learn More
Waters Corp Waters XBridge BEH Shield RP18 XP Column, 130Å, 2.5 μm
Catalog No. 50201525 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These Shield RP18 columns are designed for HPLC method development, offering alternate selectivity for phenolic compounds with their embedded carbamate group. They feature 2.5 μm eXtended Performance [XP] technology and ultra-low dispersion hardware, ensuring exceptional separation performance and robustness. The integrated Shield Technology minimizes silanol interactions, providing superior peak symmetry for basic analytes and enabling a seamless transition to UPLC.
  • Offers alternate selectivity for phenolic compounds.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path to UPLC adoption.
  • Minimizes silanol interactions with basic analytes for superior peak symmetry and efficiency.
Catalog No. 50-201-525 Supplier Waters Corp Supplier No. 186006063
These Shield RP18 columns are designed for HPLC method development, offering alternate selectivity for phenolic compounds with their embedded carbamate group. They feature 2.5 μm eXtended Performance [XP] technology and ultra-low dispersion hardware, ensuring exceptional separation performance and robustness. The integrated Shield Technology minimizes silanol interactions, providing superior peak symmetry for basic analytes and enabling a seamless transition to UPLC.
  • Offers alternate selectivity for phenolic compounds.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path to UPLC adoption.
  • Minimizes silanol interactions with basic analytes for superior peak symmetry and efficiency.