Learn More
Waters Corp Waters Column, XBridge BEH Shield RP18, 130Å, 2.5 μm, 3 mm × 50 mm
Supplier:  Waters Corp 186006057
Encompass_Preferred
XBridge BEH Shield RP18 columns provide alternate selectivity for HPLC method development, particularly for phenolic compounds, using an embedded carbamate group. These 2.5 μm XP columns feature ultra-low dispersion hardware, ensuring exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC transition.
  • Offers alternate selectivity for phenolic compounds.
  • Features embedded carbamate group in bonded phase ligand.
  • Ultra-low dispersion hardware provides exceptional separation performance.
  • Enables seamless transition to UPLC adoption.
  • Shield Technology reduces silanol interactions for improved peak shapes.
  • Complements similar C18 and C8 phase offerings.
Catalog No. 50-179-487