Learn More
Waters Corp XBridge BEH Shield RP18 XP Column, 2.1 mm X 50 mm
Catalog No. 50179222 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters XBridge BEH Shield RP18 XP Column is a (U)HPLC analytical column kit designed for reversed phase chromatography. Featuring a BEH Shield RP18 XP chemistry with C18 polar embedded, this column offers a durable stainless steel hardware construction and spherical, fully porous particles, suitable for a wide pH range of 2.0 to 11.0.
  • Designed for U(H)PLC applications and reversed phase chromatography (RP).
  • Features BEH Shield RP18 XP chemistry with C18 polar embedded.
  • Pore size of 130 Å.
  • Particle size of 2.5 μm.
  • Dimensions: 2.1 mm internal diameter and 50 mm length.
  • Made with stainless steel hardware.
  • Operates effectively in a pH range of 2.0 to 11.0.
  • Hybrid matrix with spherical, fully porous particle morphology.
Catalog No. 50-179-222 Supplier Waters Corp Supplier No. 176002563
The Waters XBridge BEH Shield RP18 XP Column is a (U)HPLC analytical column kit designed for reversed phase chromatography. Featuring a BEH Shield RP18 XP chemistry with C18 polar embedded, this column offers a durable stainless steel hardware construction and spherical, fully porous particles, suitable for a wide pH range of 2.0 to 11.0.
  • Designed for U(H)PLC applications and reversed phase chromatography (RP).
  • Features BEH Shield RP18 XP chemistry with C18 polar embedded.
  • Pore size of 130 Å.
  • Particle size of 2.5 μm.
  • Dimensions: 2.1 mm internal diameter and 50 mm length.
  • Made with stainless steel hardware.
  • Operates effectively in a pH range of 2.0 to 11.0.
  • Hybrid matrix with spherical, fully porous particle morphology.