Learn More
Waters Corp Waters XBridge BEH Shield RP18 Method Validation Kit, 130Å, 2.5 μm, 3 mm X 100 mm, 3/pk
Catalog No. 50179587 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three batches of XBridge BEH Shield RP18 columns. It is designed for HPLC method development, offering alternative selectivity and functioning effectively in a wide pH range with various solvents. It helps produce greater base separation and improved peak shapes for basic analytes.
  • Functions effectively in a wide pH range of 2 to 11.
  • Tolerates a maximum pressure of 18,000 psi (1240 Bar).
  • Utilizes patented Shield Technology for symmetrical and effective peaks.
  • Reduces silanol interactions with basic analytes for enhanced peak shape.
  • Features a 130Å pore size and 2.5 μm particle size.
  • Columns have a 3 mm inner diameter and 100 mm length.
  • Suitable for UPLC and UHPLC systems.
Catalog No. 50-179-587 Supplier Waters Corp Supplier No. 186006212
This method validation kit consists of three batches of XBridge BEH Shield RP18 columns. It is designed for HPLC method development, offering alternative selectivity and functioning effectively in a wide pH range with various solvents. It helps produce greater base separation and improved peak shapes for basic analytes.
  • Functions effectively in a wide pH range of 2 to 11.
  • Tolerates a maximum pressure of 18,000 psi (1240 Bar).
  • Utilizes patented Shield Technology for symmetrical and effective peaks.
  • Reduces silanol interactions with basic analytes for enhanced peak shape.
  • Features a 130Å pore size and 2.5 μm particle size.
  • Columns have a 3 mm inner diameter and 100 mm length.
  • Suitable for UPLC and UHPLC systems.