Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp WATERS CORPORATION XBridge BEH Shield RP18 XP Column, 130Å, 2.5 μm, 3 mm × 50 mm

Catalog No. 50179226 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XBridge BEH Shield RP18 columns provide alternate selectivity for HPLC method development, particularly for phenolic compounds, compared to straight-chain alkyl columns. These 2.5 μm eXtended Performance [XP] columns, packed in ultra-low dispersion hardware, offer exceptional separation performance, robustness, and throughput for HPLC assays, with a seamless transition path toward UPLC adoption.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 2 to 11 pH
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped: Yes
  • Bonding technology: Shield RP18
  • Silanol activity: Low
  • Particle shape: Spherical
  • Endfitting type: Parker-style
  • Surface area: 185
  • System: UPLC, UHPLC
  • Particle technology: BEH
  • USP classification: L1
  • Carbon load: 17%
  • eCord: Yes
  • UNSPSC: 41115709

Catalog No. 50-179-226 Supplier Waters Corp Supplier No. 176002567
May include imposed supplier surcharges.
Add to Cart
Add to Cart

XBridge BEH Shield RP18 columns provide alternate selectivity for HPLC method development, particularly for phenolic compounds, compared to straight-chain alkyl columns. These 2.5 μm eXtended Performance [XP] columns, packed in ultra-low dispersion hardware, offer exceptional separation performance, robustness, and throughput for HPLC assays, with a seamless transition path toward UPLC adoption.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 2 to 11 pH
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped: Yes
  • Bonding technology: Shield RP18
  • Silanol activity: Low
  • Particle shape: Spherical
  • Endfitting type: Parker-style
  • Surface area: 185
  • System: UPLC, UHPLC
  • Particle technology: BEH
  • USP classification: L1
  • Carbon load: 17%
  • eCord: Yes
  • UNSPSC: 41115709

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.