Learn More
Waters Corp Waters VanGuard Cartridge, BEH Shield RP18, 2.5 μm
Catalog No. NC2235091 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Shield RP18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream. This particular cartridge is optimized for use with all 4.6 mm I.D. XBridge BEH Shield RP18 analytical columns that contain 2.5 μm sorbent particles.
  • Extends the lifetime and performance of analytical columns.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 4.6 mm I.D. BEH Shield RP18 analytical columns.
  • Compatible with columns containing 2.5 μm sorbent particles.
Catalog No. NC2235091 Supplier Waters Corp Supplier No. 186007810
XBridge BEH Shield RP18 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination from the mobile phase stream. This particular cartridge is optimized for use with all 4.6 mm I.D. XBridge BEH Shield RP18 analytical columns that contain 2.5 μm sorbent particles.
  • Extends the lifetime and performance of analytical columns.
  • Removes particulate contamination from the mobile phase stream.
  • Optimized to protect all 4.6 mm I.D. BEH Shield RP18 analytical columns.
  • Compatible with columns containing 2.5 μm sorbent particles.