Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp HPLC Column, XBridge PST BEH130 C18, 3.5 μm, 4.6 x 100 mm, Method Validation Kit

Catalog No. 50466122 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Waters XBridge PST BEH130 C18 HPLC Column is a hybrid, reversed-phase (RP) column for exceptional reproducibility and reliable analytical methods. This Method Validation Kit includes three batches of chromatographic media from different base particles to ensure consistent performance.

  • Hybrid material
  • Inner diameter: 4.6 mm, Length: 100 mm
  • Particle size: 3.5 μm
  • Phase type: Reversed Phase (RP)
  • pH range: 1 to 12
  • Pore size: 130 Å
  • USP number: L1
  • Carbon load: 18.0%
  • Endcapping: Yes
  • Column description: XBridge
  • Package specification: C18 (RP18, ODS, Octadecyl)
  • Silanol activity: Low
  • Particle shape: Spherical
  • QC tested with selected peptide standards for consistent performance.
  • Offers outstanding batch-to-batch and column-to-column reproducibility for long-term analytical method reliability.

Catalog No. 50-466-122 Supplier Waters Corp Supplier No. 186004904
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Waters XBridge PST BEH130 C18 HPLC Column is a hybrid, reversed-phase (RP) column for exceptional reproducibility and reliable analytical methods. This Method Validation Kit includes three batches of chromatographic media from different base particles to ensure consistent performance.

  • Hybrid material
  • Inner diameter: 4.6 mm, Length: 100 mm
  • Particle size: 3.5 μm
  • Phase type: Reversed Phase (RP)
  • pH range: 1 to 12
  • Pore size: 130 Å
  • USP number: L1
  • Carbon load: 18.0%
  • Endcapping: Yes
  • Column description: XBridge
  • Package specification: C18 (RP18, ODS, Octadecyl)
  • Silanol activity: Low
  • Particle shape: Spherical
  • QC tested with selected peptide standards for consistent performance.
  • Offers outstanding batch-to-batch and column-to-column reproducibility for long-term analytical method reliability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.