Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Method Validation Kit, C18, 5 μm, 4.6 mm × 100 mm, 130 Å

Catalog No. 50466128 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This method validation kit is designed to ensure the long-term reliability and robustness of analytical methods by providing exceptional batch-to-batch and column-to-column reproducibility. It includes three batches of chromatographic media, derived from different base particles, to assess the quality, reliability, and consistency of your chromatographic method. Each batch undergoes specific QC testing with a mixture of carefully chosen peptide standards to help guarantee consistent performance.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12 pH
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped: No
  • Particle size: 5 μm
  • Pore size: 130 Å
  • QC tested with peptide standards
  • System: HPLC
  • Inner diameter: 4.6 mm
  • Length: 100 mm
  • Units per package: 3 pk

Catalog No. 50-466-128 Supplier Waters Corp Supplier No. 186005463
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This method validation kit is designed to ensure the long-term reliability and robustness of analytical methods by providing exceptional batch-to-batch and column-to-column reproducibility. It includes three batches of chromatographic media, derived from different base particles, to assess the quality, reliability, and consistency of your chromatographic method. Each batch undergoes specific QC testing with a mixture of carefully chosen peptide standards to help guarantee consistent performance.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12 pH
  • Maximum pressure: 6000 psi (415 bar)
  • Endcapped: No
  • Particle size: 5 μm
  • Pore size: 130 Å
  • QC tested with peptide standards
  • System: HPLC
  • Inner diameter: 4.6 mm
  • Length: 100 mm
  • Units per package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.