Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 130Å, 3.5 μm, 2.1 mm × 50 mm
Supplier: Waters Corp 186003563
These columns are designed and quality-control tested specifically for peptide separations. Featuring small-pore (130Å) trifunctionally bonded BEH particles, they provide a wide usable pH range, excellent low pH stability, and ultra-low column bleed. They enhance productivity and improve resolution of complex peptide digests, ensuring high-quality and repeatable test results.
- Chemistry: C18.
- Separation mode: Reversed Phase.
- Particle substrate: Hybrid.
- pH range: 1–12.
- Maximum pressure: 6000 psi (415 Bar).
- Particle size: 3.5 μm.
- Pore size: 130 Å.
- Inner diameter: 2.1 mm.
- Length: 50 mm.
- Optimal operating temperature: 20°C to 80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.