Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Peptide BEH C18 Column, 3.5 μm, 130 Å

Catalog No. 50781841 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge Peptide BEH C18 Column is optimized and QC tested for peptide separations, featuring 3.5 μm BEH particles with 130Å pore size. This column provides a wide usable pH range and superior low pH stability, making it suitable for applications in proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations.
  • Features 3.5 μm BEH particles with 130Å pore size.
  • Provides a wide usable pH range of 1 to 12.
  • Offers superior low pH stability and ultra-low column bleed.
  • Enhances resolution of complex peptide digests.
  • Improves peak shape and retention with formic acid and trifluoroacetic acid.
  • Dimensions of 1 mm inner diameter by 100 mm length.
  • Withstands a maximum pressure of 6000 psi (415 Bar).

Catalog No. 50-781-841 Supplier Waters Corp Supplier No. 186003561
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XBridge Peptide BEH C18 Column is optimized and QC tested for peptide separations, featuring 3.5 μm BEH particles with 130Å pore size. This column provides a wide usable pH range and superior low pH stability, making it suitable for applications in proteomics, protein characterization, and peptide synthesis.

  • Optimized and QC tested for peptide separations.
  • Features 3.5 μm BEH particles with 130Å pore size.
  • Provides a wide usable pH range of 1 to 12.
  • Offers superior low pH stability and ultra-low column bleed.
  • Enhances resolution of complex peptide digests.
  • Improves peak shape and retention with formic acid and trifluoroacetic acid.
  • Dimensions of 1 mm inner diameter by 100 mm length.
  • Withstands a maximum pressure of 6000 psi (415 Bar).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.