Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 130Å, 3.5 μm, 4.6 mm × 100 mm
Catalog No. 50781848 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters XBridge Peptide BEH C18 Column is a 4.6 mm × 100 mm column with 130Å pore size and 3.5 μm particle size, optimized for peptide separations. It offers wide pH stability and consistent results for proteomics and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features 130Å small-pore, trifunctionally bonded BEH particles.
  • Offers a wide usable pH range (1 to 12 pH) and superior low pH stability.
  • Ensures ultra-low column bleed.
  • Provides consistent batch-to-batch synthetic peptide and protein digest separations.
  • Utilizes advanced C18 ligand bonding for stable particle structure.
  • Delivers excellent peak shape, high efficiency, and outstanding stability.
  • Helps achieve a stable baseline for proteomics, peptide synthesis, and protein characterization.
Catalog No. 50-781-848 Supplier Waters Corp Supplier No. 186003568
Add to Cart
Add to Cart
The Waters XBridge Peptide BEH C18 Column is a 4.6 mm × 100 mm column with 130Å pore size and 3.5 μm particle size, optimized for peptide separations. It offers wide pH stability and consistent results for proteomics and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Features 130Å small-pore, trifunctionally bonded BEH particles.
  • Offers a wide usable pH range (1 to 12 pH) and superior low pH stability.
  • Ensures ultra-low column bleed.
  • Provides consistent batch-to-batch synthetic peptide and protein digest separations.
  • Utilizes advanced C18 ligand bonding for stable particle structure.
  • Delivers excellent peak shape, high efficiency, and outstanding stability.
  • Helps achieve a stable baseline for proteomics, peptide synthesis, and protein characterization.