Learn More
Waters Corp Waters XBridge Peptide Column, C18, 130Å, 3.5μm, 4.6 mm ID, 250 mm Length
Catalog No. 50781850 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, providing a wide usable pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis. Based on BEH Technology, these columns ensure excellent separation of various peptides.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) BEH particle offers wide usable pH range and superior low pH stability.
  • Delivers excellent peptide separation with narrow, symmetrical peaks.
  • Ensures method stability via special peptide map testing.
  • Features cutting-edge bonding, end-capping, and particle synthesis.
  • cGMP-certified manufacturing; includes Performance Test Chromatogram and Certification of Acceptance.
Catalog No. 50-781-850 Supplier Waters Corp Supplier No. 186003570
Add to Cart
Add to Cart
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, providing a wide usable pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis. Based on BEH Technology, these columns ensure excellent separation of various peptides.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) BEH particle offers wide usable pH range and superior low pH stability.
  • Delivers excellent peptide separation with narrow, symmetrical peaks.
  • Ensures method stability via special peptide map testing.
  • Features cutting-edge bonding, end-capping, and particle synthesis.
  • cGMP-certified manufacturing; includes Performance Test Chromatogram and Certification of Acceptance.