Learn More
Waters Corp Waters Column, C18, 130Å, 3.5 μm, 1 mm × 50 mm
Catalog No. 50781840 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is optimized and quality-controlled for peptide separation. It features trifunctionally bonded, small pore (130 Å) BEH particles, offering the widest usable pH range, improved stability at acidic pH, and extremely low bleed. It is ideal for sample quantification in proteomics, protein characterization, and peptide synthesis.
  • Optimized and quality-controlled for peptide separation.
  • Trifunctionally bonded, small pore (130 Å) BEH particles.
  • Offers the widest usable pH range.
  • Improved stability at acidic pH and extremely low bleed.
  • Ideal for sample quantification in proteomics, protein characterization, and peptide synthesis.
Catalog No. 50-781-840 Supplier Waters Corp Supplier No. 186003560
This column is optimized and quality-controlled for peptide separation. It features trifunctionally bonded, small pore (130 Å) BEH particles, offering the widest usable pH range, improved stability at acidic pH, and extremely low bleed. It is ideal for sample quantification in proteomics, protein characterization, and peptide synthesis.
  • Optimized and quality-controlled for peptide separation.
  • Trifunctionally bonded, small pore (130 Å) BEH particles.
  • Offers the widest usable pH range.
  • Improved stability at acidic pH and extremely low bleed.
  • Ideal for sample quantification in proteomics, protein characterization, and peptide synthesis.