Learn More
Waters Corp XBridge Peptide BEH C18 Column, 130Å, 3.5 μm, 2.1 mm x 250 mm
Catalog No. 50781846 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology columns are designed for peptide separations, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. These columns are suitable for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Widest usable pH range (1 to 12) and superior low pH stability.
  • Ultra-low column bleed.
  • Manufactured under cGMP and ISO 9001 standards for stable particle structure and bonding chemistry.
  • Designed for good peak form, high efficiency, and exceptional stability.
  • QC confirmed with peptide map to assure batch-to-batch consistency and reproducible performance.
Catalog No. 50-781-846 Supplier Waters Corp Supplier No. 186003566
The XBridge BEH130 C18 Peptide Separation Technology columns are designed for peptide separations, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. These columns are suitable for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Widest usable pH range (1 to 12) and superior low pH stability.
  • Ultra-low column bleed.
  • Manufactured under cGMP and ISO 9001 standards for stable particle structure and bonding chemistry.
  • Designed for good peak form, high efficiency, and exceptional stability.
  • QC confirmed with peptide map to assure batch-to-batch consistency and reproducible performance.