Learn More
Waters Corp Waters Column, C18, 3.5 μm, 130 Å, Peptide
Catalog No. 50781842 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.
Catalog No. 50-781-842 Supplier Waters Corp Supplier No. 186003562
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Small-pore (130Å) trifunctionally bonded BEH particle.
  • Widest usable pH range.
  • Superior low pH stability.
  • Ultra-low column bleed.