Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 130Å, 3.5 μm, 2.1 mm x 100 mm
Catalog No. 50781844 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns, optimized for peptide separations, feature a small-pore (130Å) trifunctionally bonded BEH particle. They provide a wide usable pH range, superior low pH stability, and ultra-low column bleed, making them suitable for proteomics, protein characterization, and peptide synthesis.
  • C18 chemistry for reversed-phase separation
  • Hybrid, spherical particles with 3.5 μm size and 130 Å pore size
  • Inner diameter of 2.1 mm and length of 100 mm
  • Wide pH range from 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Low silanol activity and endcapped
  • Molecular weight range from 1000 to 15000
  • 17% carbon load
  • QC tested for peptide applications
  • HPLC system compatible
  • USP classification L1
Catalog No. 50-781-844 Supplier Waters Corp Supplier No. 186003564
These columns, optimized for peptide separations, feature a small-pore (130Å) trifunctionally bonded BEH particle. They provide a wide usable pH range, superior low pH stability, and ultra-low column bleed, making them suitable for proteomics, protein characterization, and peptide synthesis.
  • C18 chemistry for reversed-phase separation
  • Hybrid, spherical particles with 3.5 μm size and 130 Å pore size
  • Inner diameter of 2.1 mm and length of 100 mm
  • Wide pH range from 1 to 12
  • Maximum pressure of 6000 psi (415 Bar)
  • Low silanol activity and endcapped
  • Molecular weight range from 1000 to 15000
  • 17% carbon load
  • QC tested for peptide applications
  • HPLC system compatible
  • USP classification L1