Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 5 μm, 130 Å, 2.1 mm × 150 mm
Catalog No. 50781856 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge Peptide BEH C18 columns are optimized and QC tested for peptide separations. Featuring a small-pore (130Å) trifunctionally bonded BEH particle, they offer a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed, making them ideal for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Widest usable pH range (1 to 12 pH).
  • Superior low pH stability and ultra-low column bleed.
  • Provides highest level of quality and consistency due to synthetic particles.
  • Manufactured in facilities adhering to cGMP and ISO 9001 standards.
  • Reliable for consistent synthetic peptide and protein digest separations.
Catalog No. 50-781-856 Supplier Waters Corp Supplier No. 186003576
The XBridge Peptide BEH C18 columns are optimized and QC tested for peptide separations. Featuring a small-pore (130Å) trifunctionally bonded BEH particle, they offer a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed, making them ideal for proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Widest usable pH range (1 to 12 pH).
  • Superior low pH stability and ultra-low column bleed.
  • Provides highest level of quality and consistency due to synthetic particles.
  • Manufactured in facilities adhering to cGMP and ISO 9001 standards.
  • Reliable for consistent synthetic peptide and protein digest separations.