Learn More
Waters Corp Waters Column, Peptide BEH C18, 130 Å, 5 μm, 1 mm × 150 mm
Catalog No. 50781853 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH130 C18 Peptide Separation Technology (PST) columns are designed for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. This makes them ideal for proteomics, protein characterization, and peptide synthesis applications.
  • QC tested for peptide separations.
  • Utilizes a small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers a wide usable pH range of 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Suitable for proteomics, protein characterization, and peptide synthesis.
  • Withstands pressures up to 6000 psi (415 Bar).
  • Recommended operating temperature between 20°C to 80°C.
  • Features a hybrid, spherical particle substrate.
Catalog No. 50-781-853 Supplier Waters Corp Supplier No. 186003573
XBridge BEH130 C18 Peptide Separation Technology (PST) columns are designed for peptide separations. They feature a small-pore (130Å) trifunctionally bonded BEH particle, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. This makes them ideal for proteomics, protein characterization, and peptide synthesis applications.
  • QC tested for peptide separations.
  • Utilizes a small-pore (130Å) trifunctionally bonded BEH particle.
  • Offers a wide usable pH range of 1 to 12.
  • Provides superior low pH stability and ultra-low column bleed.
  • Suitable for proteomics, protein characterization, and peptide synthesis.
  • Withstands pressures up to 6000 psi (415 Bar).
  • Recommended operating temperature between 20°C to 80°C.
  • Features a hybrid, spherical particle substrate.