Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XBridge Column, C18, 5 μm, 130 Å, 4.6 mm x 50 mm

Catalog No. 50781858 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These columns are optimized and QC tested for peptide separations. The 130Å trifunctionally bonded BEH particle provides a wide pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid, spherical shape
  • pH range: 1 to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped with low silanol activity
  • Molecular weight range: 1000 to 15000
  • Particle size: 5 μm; Pore size: 130 Å
  • QC tested for peptide applications
  • HPLC system column format
  • Surface area: 185; Carbon load: 17%
  • USP classification: L1

Catalog No. 50-781-858 Supplier Waters Corp Supplier No. 186003578
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These columns are optimized and QC tested for peptide separations. The 130Å trifunctionally bonded BEH particle provides a wide pH range, superior low pH stability, and ultra-low column bleed for proteomics, protein characterization, and peptide synthesis.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid, spherical shape
  • pH range: 1 to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped with low silanol activity
  • Molecular weight range: 1000 to 15000
  • Particle size: 5 μm; Pore size: 130 Å
  • QC tested for peptide applications
  • HPLC system column format
  • Surface area: 185; Carbon load: 17%
  • USP classification: L1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.