Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge Peptide BEH C18 Column, 5 μm, 130Å, 4.6 mm X 250 mm

Catalog No. 50781861 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations, offering the widest usable pH range, superior low pH stability, and ultra-low column bleed. They are designed for assaying samples in proteomics, protein characterization, and peptide synthesis, providing excellent separation and superior peak shape. These columns allow for seamless method migration from sub-2 μm UPLC Technology to 10 μm preparative HPLC separations.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Molecular weight range: 1,000 to 15,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 130 Å
  • Inner diameter: 4.6 mm
  • Length: 250 mm

Catalog No. 50-781-861 Supplier Waters Corp Supplier No. 186003581
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations, offering the widest usable pH range, superior low pH stability, and ultra-low column bleed. They are designed for assaying samples in proteomics, protein characterization, and peptide synthesis, providing excellent separation and superior peak shape. These columns allow for seamless method migration from sub-2 μm UPLC Technology to 10 μm preparative HPLC separations.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12 pH
  • Maximum pressure: 6000 psi (415 Bar)
  • Molecular weight range: 1,000 to 15,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 130 Å
  • Inner diameter: 4.6 mm
  • Length: 250 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.