Learn More
Waters Corp Waters Column, XBridge BEH C18, Peptide, 130Å 5 μm
Catalog No. 50781860 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XBridge BEH130 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. They offer a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed, making them ideal for proteomics, protein characterization, and peptide synthesis applications.
  • Optimized and QC tested for peptide separations.
  • Wide usable pH range (1-12) with superior low pH stability and ultra-low column bleed.
  • Utilizes BEH technology and synthetic particles for consistent performance.
  • Manufactured in a cGMP, ISO 9002-certified facility.
  • Each column includes a Performance Test Chromatogram and Certification of Approval.
Catalog No. 50-781-860 Supplier Waters Corp Supplier No. 186003580
These XBridge BEH130 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. They offer a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed, making them ideal for proteomics, protein characterization, and peptide synthesis applications.
  • Optimized and QC tested for peptide separations.
  • Wide usable pH range (1-12) with superior low pH stability and ultra-low column bleed.
  • Utilizes BEH technology and synthetic particles for consistent performance.
  • Manufactured in a cGMP, ISO 9002-certified facility.
  • Each column includes a Performance Test Chromatogram and Certification of Approval.