Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 130Å, 5 μm, 2.1 mm x 250 mm
Catalog No. 50781857 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for assay samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Widest usable pH range (1–12 pH) with superior low pH stability and ultra-low column bleed.
  • Trifunctionally bound BEH particle supports peptide synthesis and protein characterization.
  • Produced in a cGMP, ISO 9002 certified facility using ultra-pure reagents for peak shape, efficiency, and stability.
  • Each batch qualified with a peptide separation to ensure repeatable performance.
  • Packaged with a Performance Test Chromatogram and Certification of Acceptance.
Catalog No. 50-781-857 Supplier Waters Corp Supplier No. 186003577
Add to Cart
Add to Cart
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed for assay samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations.
  • Widest usable pH range (1–12 pH) with superior low pH stability and ultra-low column bleed.
  • Trifunctionally bound BEH particle supports peptide synthesis and protein characterization.
  • Produced in a cGMP, ISO 9002 certified facility using ultra-pure reagents for peak shape, efficiency, and stability.
  • Each batch qualified with a peptide separation to ensure repeatable performance.
  • Packaged with a Performance Test Chromatogram and Certification of Acceptance.