Learn More
Waters Corp XBridge Peptide BEH C18 Prep Guard Cartridge, 130Å, 10 µm, 19 mm X 10 mm, 1/pk
Supplier:  Waters Corp 186004464
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Catalog No. 50-466-005