Learn More
Waters Corp HPLC Guard Cartridge, BEH C18, 130Å, 5μm, 19 x 10 mm
Supplier: Waters Corp 186004468
This guard column is designed to extend the lifespan of preparative columns. The packing material undergoes peptide digest QC test criteria, ensuring consistent performance.
- Extended column life: Prolongs the life of preparative columns.
- Quality assurance: Packing material meets peptide digest QC test criteria for consistent performance.
- Reversed phase mode: Operates in RP mode, suitable for a wide range of chromatographic applications.
- Wide pH stability: Broad pH range of 1–12 for versatile mobile phase conditions.
- Specific pore size: 130 Å pore size optimized for peptide applications.
- C18 chemistry: Utilizes C18 (Octadecyl) chemistry, a robust stationary phase for hydrophobic interactions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.