Learn More
Waters Corp XBridge Peptide BEH C18 Column, 130Å, 10 µm, 4.6 mm X 100 mm, 1/pk
Catalog No. 50781921 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Catalog No. 50-781-921 Supplier Waters Corp Supplier No. 186003649
The XBridge BEH130 C18 Peptide Separation Technology columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.