Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation OBD Prep Column, C18, 130 Å, 5 μm, 19 mm x 50 mm

Catalog No. 50781866 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This preparative column, designed for peptide separation in research and development, provides consistent chromatographic tools for analysis and isolation. It utilizes C18 hybrid particles, ensuring sharp, symmetrical peaks across a wide range of peptide properties.

  • Reversed phase separation mode
  • Hybrid, spherical particle shape
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped with low silanol activity
  • Particle size: 5 μm
  • Pore size: 130 Å
  • QC tested for peptide applications
  • Format: OBD prep column
  • System: HPLC
  • USP classification: L1
  • Dimensions: 19 mm inner diameter, 50 mm length
  • Carbon load: 18%
  • Provides uniform, optimally packed beds
  • Ensures excellent column lifetime
  • Offers direct scalability from analytical to preparative scales

Catalog No. 50-781-866 Supplier Waters Corp Supplier No. 186003586
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This preparative column, designed for peptide separation in research and development, provides consistent chromatographic tools for analysis and isolation. It utilizes C18 hybrid particles, ensuring sharp, symmetrical peaks across a wide range of peptide properties.

  • Reversed phase separation mode
  • Hybrid, spherical particle shape
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 Bar)
  • Endcapped with low silanol activity
  • Particle size: 5 μm
  • Pore size: 130 Å
  • QC tested for peptide applications
  • Format: OBD prep column
  • System: HPLC
  • USP classification: L1
  • Dimensions: 19 mm inner diameter, 50 mm length
  • Carbon load: 18%
  • Provides uniform, optimally packed beds
  • Ensures excellent column lifetime
  • Offers direct scalability from analytical to preparative scales

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.