Learn More
Waters Corp Waters Corporation Prep Column, C18, 130Å, 10 μm, 19 mm X 250 mm
Catalog No. 50781930 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH130 C18 Peptide Separation Technology (PST) preparative columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Optimized and QC tested for peptide separations
  • Small-pore (130Å) trifunctionally bonded BEH particle
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed
  • Optimum Bed Density (OBD) hardware format for superior column lifetime
Catalog No. 50-781-930 Supplier Waters Corp Supplier No. 186003658
Add to Cart
Add to Cart
Request a Quote
The XBridge BEH130 C18 Peptide Separation Technology (PST) preparative columns are optimized and QC tested for peptide separations. The small-pore (130Å) trifunctionally bonded BEH particle offers the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis. The Optimum Bed Density (OBD) hardware format ensures superior column lifetime under a wide variety of operating conditions.
  • Optimized and QC tested for peptide separations
  • Small-pore (130Å) trifunctionally bonded BEH particle
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed
  • Optimum Bed Density (OBD) hardware format for superior column lifetime