Learn More
Waters Corp XBridge Peptide BEH C18 Prep Column, 30 mm x 50 mm
Catalog No. 50781931 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These preparative columns are designed for peptide isolations and separations. They feature small-pore (130Å) trifunctionally bonded BEH particles, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. They are suitable for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized for peptide separations and QC tested.
  • Trifunctionally bonded BEH particles (130Å pore) for diverse separation selectivities.
  • Wide usable pH range (1 to 12) and superior low pH stability.
  • Ultra-low column bleed.
  • Optimum Bed Density (OBD) hardware ensures superior column lifetime.
  • Outstanding peak capacity and shape in TFA or FA ion pair eluents, outperforming 100% silica-based C18 columns.
  • Each batch QC tested with complex protein digest for reproducible performance.
Catalog No. 50-781-931 Supplier Waters Corp Supplier No. 186003659
Add to Cart
Add to Cart
Request a Quote
These preparative columns are designed for peptide isolations and separations. They feature small-pore (130Å) trifunctionally bonded BEH particles, offering a wide usable pH range, superior low pH stability, and ultra-low column bleed. They are suitable for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized for peptide separations and QC tested.
  • Trifunctionally bonded BEH particles (130Å pore) for diverse separation selectivities.
  • Wide usable pH range (1 to 12) and superior low pH stability.
  • Ultra-low column bleed.
  • Optimum Bed Density (OBD) hardware ensures superior column lifetime.
  • Outstanding peak capacity and shape in TFA or FA ion pair eluents, outperforming 100% silica-based C18 columns.
  • Each batch QC tested with complex protein digest for reproducible performance.