Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation HPLC Column, Peptide BEH C18, 3.5 μm, 300 Å

Catalog No. 50466123 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This method validation kit provides exceptional batch-to-batch and column-to-column reproducibility, crucial for long-term reliability and robustness of analytical methods. It includes three batches of chromatographic media, derived from different base particles, to assess the quality, reliability, and consistency of your chromatographic method. Each batch of the BEH300 C18, 3.5μm chemistry is specifically QC tested with a mixture of carefully selected peptide standards to help ensure consistent performance.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 bar)
  • Particle shape: Spherical
  • Particle size: 3.5 μm
  • Pore size: 300 Å
  • QC tested: Peptide
  • System: HPLC
  • Inner diameter: 4.6 mm
  • Length: 100 mm
  • Units per package: 3 pk

Catalog No. 50-466-123 Supplier Waters Corp Supplier No. 186004905
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This method validation kit provides exceptional batch-to-batch and column-to-column reproducibility, crucial for long-term reliability and robustness of analytical methods. It includes three batches of chromatographic media, derived from different base particles, to assess the quality, reliability, and consistency of your chromatographic method. Each batch of the BEH300 C18, 3.5μm chemistry is specifically QC tested with a mixture of carefully selected peptide standards to help ensure consistent performance.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Hybrid
  • pH range: 1 to 12
  • Maximum pressure: 6000 psi (415 bar)
  • Particle shape: Spherical
  • Particle size: 3.5 μm
  • Pore size: 300 Å
  • QC tested: Peptide
  • System: HPLC
  • Inner diameter: 4.6 mm
  • Length: 100 mm
  • Units per package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.