Learn More
Waters Corp Waters XBridge Peptide BEH C18 Column, 300Å, 3.5 μm, 2.1 mm, 100 mm
Catalog No. 50781884 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XBridge Peptide BEH C18 Column, 300Å, 3.5 μm, 2.1 mm, 100 mm
Catalog No. 50-781-884 Supplier Waters Corp Supplier No. 186003608
Waters XBridge Peptide BEH C18 Column, 300Å, 3.5 μm, 2.1 mm, 100 mm