Learn More
Waters Corp Waters Corporation Column, C18, 300Å, 3.5 μm, 4.6 mm X 50 mm
Catalog No. 50781887 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, offering the widest usable pH range, superior low pH stability, and ultra-low column bleed. These columns are designed for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations
  • Wide-pore (300Å) trifunctionally bonded BEH particle
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed
Catalog No. 50-781-887 Supplier Waters Corp Supplier No. 186003611
The XBridge BEH300 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations. They feature a wide-pore (300Å) trifunctionally bonded BEH particle, offering the widest usable pH range, superior low pH stability, and ultra-low column bleed. These columns are designed for assaying samples in proteomics, protein characterization, and peptide synthesis.
  • Optimized and QC tested for peptide separations
  • Wide-pore (300Å) trifunctionally bonded BEH particle
  • Widest usable pH range
  • Superior low pH stability
  • Ultra-low column bleed